Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (H, L) Rabbit anti-Dog, Polyclonal , Abnova™

Catalog No. 89959191 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-959-191 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-959-191 Supplier Abnova Corporation Supplier No. PAB9181
Add to Cart
Add to Cart

Rabbit anti-dog IgG recognizes dog IgG whole molecule.

Specifications

Antigen IgG (H, L)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit anti-dog IgG recognizes dog IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography.
Dilution The optimal working dilution should be determined by the end user.
Formulation Liquid
Host Species Rabbit
Immunogen Dog IgG whole molecule
Purification Method This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Dog
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.