Learn More
Description
Specifications
Specifications
| Antigen | IgG (H&L) |
| Applications | Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | TRITC |
| Host Species | Rabbit |
| Immunogen | Human IgG whole molecule |
| Quantity | 1 mL |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Content And Storage | Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.