Learn More
Specifications
Specifications
| Antigen | IgG (H, L) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit anti-rat IgG recognizes rat IgG whole molecule. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | In 10mM sodium phosphate, 150mM NaCl, pH 7.2 (0.01% Sodium Azide). |
| Host Species | Rabbit |
| Immunogen | Rat IgG whole molecule |
| Purification Method | This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against rat IgG and rat serum. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.