Learn More
AdipoGen IgG h mAb rec Multiclonal
SDP
Supplier:  AdipoGen AG27B6300C100
Encompass_Preferred
Recombinant Antibody. Recognizes different epitopes on all four different subclasses of human IgG. Applications ELISA, FACS, IHC, WB. Clone . Isotype Mouse IgG2a. Formulation Liquid. In PBS. Multiclonals are recombinant secondary antibodies that combine the best of polyclonal antisera and hybridoma monoclonal antibodies, while eliminating their disadvantages, plus they add the quality of recombinant reagents. Multiclonal antibodies consist of carefully adjusted mixtures recognizing different epitopes on all four different subclasses of human IgG. Their respective epitope binding sites do not compete with each other, therefore amplifying signal strengths. Multiclonals can contain up to 17 different individually tested monoclonal recombinant antibodies.
Catalog No. 50-256-3145