Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgG (Human) ELISA Kit

Catalog No. 89101113 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
96 Assays
Sample Type:
Cell Culture, Milk, Plasma, Saliva, Serum, Urine
Plasma, Serum
Sample Volume:
100 μL
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sample Type Sample Volume
89-101-113 1 Kit Cell Culture, Milk, Plasma, Saliva, Serum, Urine 50 μL
89-028-465 96 Assays Plasma, Serum 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-113 Supplier Abnova™ Supplier No. KA3817
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IgG

  • Sample Type: Plasma, Serum
  • Sample Volume: 100μL

Quantification

Specifications

Calibration Range 0.625 to 40.0ng/mL
Cross Reactivity Human
Description IgG (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IgG.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Cell Culture, Milk, Plasma, Saliva, Serum, Urine
Sample Volume 50 μL
Content And Storage Store SP conjugate and biotinylated antibody and standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type IgG (Human) ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.