Learn More
AdipoGen IgG human pAb HRP
SDP
Supplier:  AdipoGen AG29B0005ER050
Encompass_Preferred
Polyclonal Antibody. Reacts with the heavy and light chains of human IgG. Shows minimal cross-reactivity with IgG from other species. Source Goat. Applications IHC, WB. Liquid. In PBS containing 50% glycerol and 0.02% sodium azide. The anti-IgG human, pAb HRP is cross-adsorbed against human IgM and IgA and affinity-purified on pooled human IgG covalently linked to agarose.
Catalog No. 50-256-3103