Learn More
Apexbio Technology LLC IgG light chain variable region [Homo sapiens]/IgM/kappa antibody [Mus musculus] 5mg
SDP
Catalog No. 5000096432
Encompass_Preferred
IgG light chain variable region Homo sapiens IgM kappa antibody Mus musculus is an antibody fragment associated with antigen recognition The light chain variable region (VL) is encoded by V and J segments on chromosome 2 (IGK locus) and is essential for antibody-antigen interactions IgM antibodies including the murine kappa type are expressed as membrane-bound monomers on B lymphocytes and form pentamers in circulation to enhance antigen binding during early humoral responses Alterations in and light chain ratios are used as diagnostic markers for B-cell malignancies Free immunoglobulin light chains are implicated as mediators and biomarkers in inflammatory research due to their activation of mast cells neutrophils and sensory neurons
Catalog No. 50-000-96432 Supplier Apexbio Technology LLC Supplier No. A10705
IgG light chain variable region Homo sapiens IgM kappa antibody Mus musculus is an antibody fragment associated with antigen recognition The light chain variable region (VL) is encoded by V and J segments on chromosome 2 (IGK locus) and is essential for antibody-antigen interactions IgM antibodies including the murine kappa type are expressed as membrane-bound monomers on B lymphocytes and form pentamers in circulation to enhance antigen binding during early humoral responses Alterations in and light chain ratios are used as diagnostic markers for B-cell malignancies Free immunoglobulin light chains are implicated as mediators and biomarkers in inflammatory research due to their activation of mast cells neutrophils and sensory neurons