Learn More
Alpco Ltd Mouse IgG ELISA
SDP
Supplier:  Alpco Ltd 41IGGMSE01
Encompass
The Mouse IgG ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgG in biological samples of mice. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3593