Learn More
AdipoGen IgG mouse pAb HRP
SDP
Supplier:  AdipoGen AG29B0003ER050
Encompass_Preferred
Polyclonal Antibody. Reacts with the heavy and light chains of mouse IgG1. Shows minimal cross-reactivity with human immunoglobulins. Source Goat. Applications ELISA, IHC, WB. Liquid. In PBS containing 50% glycerol. The anti-IgG1 mouse, pAb HRP is cross-adsorbed against mouse IgM, IgG2a, IgG2b, IgG3 and IgA, pooled human sera and purified human immunoglobulins.
Catalog No. 50-256-3106