Learn More
AdipoGen IgG mouse pAb HRP
SDP
Catalog No. 502563105
Encompass_Preferred
Polyclonal Antibody. Reacts with mouse IgG1, IgG2a, IgG2b and IgG3. Source Goat. Applications IHC, WB. Liquid. In PBS containing 50% glycerol. The anti-IgG mouse, pAb HRP is cross-absorbed against mouse IgM and IgA, pooled human sera and purified human paraproteins and affinity-purified on pooled mouse IgG covalently linked to agarose.
Catalog No. 50-256-3105 Supplier AdipoGen Supplier No. AG29B0004ER050
Polyclonal Antibody. Reacts with mouse IgG1, IgG2a, IgG2b and IgG3. Source Goat. Applications IHC, WB. Liquid. In PBS containing 50% glycerol. The anti-IgG mouse, pAb HRP is cross-absorbed against mouse IgM and IgA, pooled human sera and purified human paraproteins and affinity-purified on pooled mouse IgG covalently linked to agarose.