Learn More
QA Bio Inc IgG N-glycan library, unlabelled, 25 μg
Catalog No. 502236573
Encompass
A mixture of fucosylated, bi-antennary glycan standards with variable sialylation released from human IgG antibody glycoprotein. • 25 µgs
This item is not returnable. View return policy
Catalog No. 50-223-6573 Supplier QA Bio Inc Supplier No. CLIBNIGG01
A mixture of fucosylated, bi-antennary glycan standards with variable sialylation released from human IgG antibody glycoprotein. • 25 µgs