Learn More
Alpco Ltd Rat IgG ELISA
SDP
Supplier:  Alpco Ltd 41IGGRTE01
Encompass
The Rat IgE ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgE in the serum or plasma of rats. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3594