Learn More
AdipoGen IgG rat pAb HRP
SDP
Supplier:  AdipoGen AG29B0001ER050
Encompass_Preferred
Polyclonal Antibody. Reacts with the heavy and light chains of rat IgG and with the light chains of rat IgM. Shows minimal cross-reactivity with mouse immunoglobulins. Source Goat. Applications IHC, WB. Liquid. In PBS containing 50% glycerol. The anti-IgG rat, pAb HRP is cross-adsorbed against pooled mouse sera and pooled mouse plasmacytoma/hybridoma proteins and affinity-purified on pooled rat IgG covalently linked to agarose.
Catalog No. 50-256-3108