Learn More
AdipoGen IgG rat pAb R-PE
SDP
Supplier:  AdipoGen AG29B0001RC050
Encompass_Preferred
Polyclonal Antibody. Reacts with the heavy and light chains of rat IgG, and with the light chains of rat IgM. Shows minimal cross-reactivity with mouse immunoglobulins. Source Goat. Applications FACS, ICC. Liquid. In PBS containing 0.02% sodium azide. The anti-IgG rat, pAb R-PE is cross-adsorbed against pooled mouse sera and pooled mouse plasmacytoma/hybridoma proteins and affinity-purified on pooled rat IgG covalently linked to agarose.
Catalog No. 50-256-3107