Learn More
Bioworld IGG SEPAROPOREAGAROSE)- F
Catalog No. 50197694
Encompass
IGG SEPAROPORE (AGAROSE)- FUSION KIT: A complete kit to bind Protein A fusion proteins expressed in E. Coli or yeast. Consists of affinity chromatography gel matrix, IgG-Separopore(4% agarose beads), including all the components/bufferst...
Catalog No. 50-197-694 Supplier Bioworld Supplier No. 201810141
IGG SEPAROPORE (AGAROSE)- FUSION KIT: A complete kit to bind Protein A fusion proteins expressed in E. Coli or yeast. Consists of affinity chromatography gel matrix, IgG-Separopore(4% agarose beads), including all the components/bufferst...