Learn More
Med Vet International Bovine IgG Supplement 57.5 Natural Bovine Dried Colostrum, 320gm
Supplier: Med Vet International PHIGG575320GM
Bovine IgG Supplement 57.5 is made from natural bovine colostrum and is used to replace or supplement maternal colostrum as an aid in the treatment of Failure of Passive Transfer (FPT) in newborn calves. ∙ 15% higher Bovine IgG content than most other leading brands of supplements & replacers on the market ∙ Actual colostrum, no other ingredients added, nothing taken away ∙ Not a generic ∙ Easy to use, mixes instantly ∙ Each bag of Supplement 57.5 provides > 57.5 g of Bovine IgG (> 18% IgG) ∙ USDA approved and licensed ∙ Guaranteed for safety, efficacy, purity and potency. ∙ High energy/brown fat metabolism from colostral fat. ∙ Made from large pools of colostrum from many different individuals which guarantees product contains a wide variety of antibodies to all agents in the dairy calf environment including protective levels to E. coli