Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG (Whole Molecule) Goat anti-Human , FITC, Secondary Antibody, MP Biomedicals™

Catalog No. MP0855197 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0855197 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0855197 Supplier MP Biomedicals, Inc Supplier No. 0855197
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

  • It is suitable for use in direct immunofluorescence, as well as for ANA (Anti-Nuclear Antibody)-indirect immunofluorescence.
  • The antibody may also be used in FACS-based assays.
  • It is also suitable for use as a reagent in immunofluorescence assays (IFA), cell staining (fluorescent microscopy and cell sorting), immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections, and blot immunostaining.
  • Use of these affinity purified antibodies is recommended to avoid non-specific binding from the inherent antibodies of host animals.
  • (Note: F(ab')2 fragments are recommended for staining of cells or tissues which contain Fc receptors). Fluorescein-Conjugated Goat Affinity Purified Antibody to Human IgG (Whole Molecule) is fluorescein-5-isothiocyanate (FITC “Isomer I”)-conjugated goat affinity purified antibody to human IgG (whole molecule) and buffer salts.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunoassay
Classification Polyclonal
Concentration Total Protein: 1.12mg/mL; Active Antibody: 1.0mg/mL
Conjugate FITC
Description Fluorescein-Conjugatedgoat Affinity Purified Antibody to Human IgG (Whole Molecule) is fluorescein-5-isothiocyanate (FITC “Isomer I”)-conjugatedgoat affinity purified antibody to human IgG (whole molecule) and buffer salts.
Host Species Goat
Immunogen Pooled normal human serum
Purification Method Affinity Purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Content And Storage Store unopened product at 2-8°C. Store opened material no longer than 4 weeks at 2-8°C. For long term storage, aliquot and store with 40%glycerol (v/v) at -20°C.
Form Liquid
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.