Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Antibody (RM106), Biotin, Novus Biologicals™
SDP

Catalog No. NBP262006B
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262006B 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262006B Supplier R&D Systems Supplier No. NBP262006B

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for ELISA, Western Blot.

Specifications

Antigen IgG1
Applications ELISA, Western Blot
Classification Monoclonal
Clone RM106
Concentration 1 mg/mL
Conjugate Biotin
Dilution Western Blot 0.1 ug/ml - 0.5 ug/ml, Flow Cytometry, ELISA 0.005 ug/ml-0.2 ug/ml, Immunohistochemistry 0.5 - 2 ug/mL, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunocytochemistry 0.5 - 2 ug/mL
Formulation PBS, 50% Glycerol, 1% BSA with 0.09% Sodium Azide
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Rabbit
Immunogen Mouse IgG
Purification Method Protein A purified
Quantity 50 μg
Primary or Secondary Secondary
Test Specificity This antibody reacts to the Fc region of mouse IgG1. No cross reactivity with mouse IgG2a, IgG3, IgM, IgA, IgE, human IgG, or rat IgG. It may slightly cross react to mouse IgG2b, and may also react to goat IgG.
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.