Learn More
Athens Research And Technology Inc IGG1 HU MYELOMA PLASMA 5MG
Supplier:  Athens Research And Technology Inc 16160907071M5MG
Encompass_Preferred
Immunoglobulin G1, Human Myeloma Plasma: 5x1 MG; Frozen in 20 mM phosphate, pH 7.4, with 150 mM NaCl and 0.05% NaN3; MW 146,000; EC=1.36; Purity: > =95% by SDS-PAGE. Native; Purified from human myeloma plasma in the Athens Research & Tech lab.
Catalog No. 50-489-800