Learn More
Medchemexpress LLC IgG1 Human CHO C1 500ug
SDP
Supplier:  Medchemexpress LLC HYP73903500UG
Encompass_Preferred
The IgG1 protein is the constant region of the immunoglobulin heavy chain and serves as a receptor during the recognition phase of humoral immunity It triggers clonal expansion and differentiation of B lymphocytes into immunoglobulin-secreting plasma cells IgG1 Protein Human (CHO C103S) is the recombinant human-derived IgG1 protein expressed by CHO with tag free
Catalog No. 50-001-79336