Learn More
AdipoGen IgG1 human mAb Biotin
SDP
Supplier:  AdipoGen AG29B0007BC050
Encompass_Preferred
Monoclonal Antibody. Recognizes human IgG1Fc. Isotype Mouse IgG1. Clone Huf 5.4. Applications ELISA, FACS, WB. Liquid. In PBS sterile containing 0.1% sodium azide. Detects human IgG1Fc proteins and can be used as a secondary control antibody.
Catalog No. 50-256-3099