Learn More
Enzo Life Sciences Mouse IgG1 isotype control, monoclonal antibody (MOPC-21) (DyLight 488 conjugate) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISAB600488F
Encompass_Preferred
Clone: MOPC-21. Host: Mouse. Isotype: IgG1. Source: Purified from ascites. Species Reactivity: Species independent. Applications: Flow Cytometry. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-667-393