Learn More
Sigma Aldrich Fine Chemicals Biosciences IgG1 Lambda from human mye1MG
Catalog No. 502904599
Encompass
IgG is the main antibody type found in plasma and extracellular fluid and is expressed on B cell membrane. IgG is further subdivided into four subclass- IgG1 IgG2 IgG3 and IgG4.Elevation in antigen specific IgG1 is associated with symptomatic infection and Th2-regulation. IgG1» from human myeloma can be used in quantitative ELISA.Human IgG1» react specifically with human IgG.
Catalog No. 50-290-4599 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I50291MG
IgG is the main antibody type found in plasma and extracellular fluid and is expressed on B cell membrane. IgG is further subdivided into four subclass- IgG1 IgG2 IgG3 and IgG4.Elevation in antigen specific IgG1 is associated with symptomatic infection and Th2-regulation. IgG1» from human myeloma can be used in quantitative ELISA.Human IgG1» react specifically with human IgG.