Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Mouse anti-Human, HRP, Secondary Antibody, MP Biomedicals™

Catalog No. MP08634521 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08634521 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08634521 Supplier MP Biomedicals, Inc Supplier No. 08634521
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Anti-Human IgG1 Secondary Antibody

TRUSTED_SUSTAINABILITY

Specifications

Applications ELISA, EIA
Classification Monoclonal
Clone HP6069
Conjugate HRP
Description Mouse Anti-Human IgG1
Host Species Mouse
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.