Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Mouse anti-Rat, AF 488, Clone: G11C5, Southern Biotech™
SDP

Catalog No. OB306130 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB306130 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB306130 Supplier Southern Biotechnology Associates Supplier No. 306130
Add to Cart
Add to Cart

Mouse Monoclonal Secondary Antibody

SouthernBiotech monoclonal secondary antibodies offer high sensitivity and low background for use in a variety of applications.

Specifications

Antigen IgG1
Applications Flow Cytometry, ELISA, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone G11C5
Concentration 0.5mg/mL
Conjugate Alexa Fluor 488
Formulation Supplied in PBS/NaN3
Host Species Mouse
Immunogen Rat IgG1 hybridoma
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 2-8°C; Protect from light
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.