Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IgG1 Protein Human 500ug
SDP

Catalog No. 5000197004
Encompass_Preferred

Recombinant human IgG1 protein (D239E, L241E) is the Fc constant region expressed in HEK293 cells and supplied tag-free. It is intended for use in immunology and biochemical assays that require the human IgG1 Fc domain, and can support studies of Fc receptor binding and assay development.

  • Recombinant human IgG1 Fc region with D239E and L241E mutations.
  • Expressed in HEK293 mammalian cells.
  • Tag free format for native Fc studies.
  • Suitable for immunology and biochemical assay development.
  • Useful for Fc receptor interaction and binding studies.

Catalog No. 50-001-97004 Supplier Medchemexpress LLC Supplier No. HYP70796A500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human IgG1 protein (D239E, L241E) is the Fc constant region expressed in HEK293 cells and supplied tag-free. It is intended for use in immunology and biochemical assays that require the human IgG1 Fc domain, and can support studies of Fc receptor binding and assay development.

  • Recombinant human IgG1 Fc region with D239E and L241E mutations.
  • Expressed in HEK293 mammalian cells.
  • Tag free format for native Fc studies.
  • Suitable for immunology and biochemical assay development.
  • Useful for Fc receptor interaction and binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.