Learn More
Sino Biological Recombinant Human IgG1-Fc Protein (C103S), HPLC-verified
Supplier:  Sino Biological 10702HNAH1
Encompass_Preferred
A DNA sequence encoding the human IgG1 Fc region (P01857-1) (Glu99-Lys330) (one aa mutation, 103 Cys/Ser) was expressed.
Catalog No. 50-211-6355