Learn More
Sino Biological Human IgG1-Fc Protein (103 Cys/Ser), Biotinylated 100ug
Supplier:  Sino Biological 10702HNAHB100
Encompass_Preferred
A DNA sequence encoding the IGHG1 (AAC82527.1) (Glu99-Lys330) (one aa mutation,103 Cys/Ser) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6173