Learn More
Abcam Native Human IgG1 protein, 1MG
Catalog No. NC0104062
Encompass_Preferred
Native Human IgG1 protein is a Human Full Length IgG1 protein with >95% purity and suitable for SDS-PAGE and ELISA. The predicted molecular weight of ab90283 native protein is 146 kDa. - Save time and ensure accurate result-use our native IgG1 protein as an isotype control - No cross-reaction from other immunoglobulin isotypes
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant gamma 1, Ig gamma-1 chain C region, Ig gamma-1 chain C region EU, Ig gamma-1 chain C region KOL, Ig gamma-1 chain C region NIE, IGHG1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0104062 Supplier Abcam Supplier No. AB902831MG
Native Human IgG1 protein is a Human Full Length IgG1 protein with >95% purity and suitable for SDS-PAGE and ELISA. The predicted molecular weight of ab90283 native protein is 146 kDa. - Save time and ensure accurate result-use our native IgG1 protein as an isotype control - No cross-reaction from other immunoglobulin isotypes
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin heavy constant gamma 1, Ig gamma-1 chain C region, Ig gamma-1 chain C region EU, Ig gamma-1 chain C region KOL, Ig gamma-1 chain C region NIE, IGHG1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement