Learn More
ACROBiosystems ACROBiosystems Human IgG1 Fc Protein, Flag Tag
Supplier:  ACROBiosystems IG1H52C9500UG
Encompass_Preferred
Recombinant Protein;500UG;Human IgG1 Fc, Flag Tag (IG1-H52C9) is expressed from human 293 cells (HEK293). It contains AA Pro 100 - Lys 330 (Accession # P01857-1 (C103S)).;This protein carries a flag tag at the C-terminus. The protein has a calculated MW of 27.1 kDa. The protein migrates as 28-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6533