Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human IgG1 Fc protein, Avi Tag (Avitag™);HEK293;500UG;IG1,IgG1 Fc,IgG2,IgG3,IgG4,IgG2a,IgG Fc,IgG1
Supplier:  ACROBiosystems IG1H8213500UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human IgG1 Fc protein, Avi Tag (Avitag™);500UG;Biotinylated Human IgG Fc, Avi Tag (IG1-H8213) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession # P01857-1 ).;This protein carries an Avi tag at the C-terminus. The protein has a calculated MW of 28.4 kDa. The protein migrates as 36-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Recombinant Protein G (Cat. No. RPG-S3140) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IgG1 Fc, Avi Tag (Cat. No. IG1-H8213) with a linear range of 13-512 ng/mL (Routinely tested).
Catalog No. 50-165-8314