Learn More
ACROBiosystems Recombinant Protein;Human IgG1 Fc Protein, His Tag;HEK293;1MG;IG1,IgG1 Fc,IgG2,IgG3,IgG4,IgG2a,IgG Fc,IgG1
Supplier:  ACROBiosystems IG1H52251MG
Encompass_Preferred
Recombinant Protein;Human IgG1 Fc Protein, His Tag;1MG;Human IgG Fc, His Tag (IG1-H5225) is expressed from human 293 cells (HEK293). It contains AA Glu 99 - Lys 330 (Accession # P01857).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27 kDa. The protein migrates as 34 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8915