Learn More
Cayman Chemical IgG1humn ELISA Stndard 1ea
SDP
Catalog No. 502945791
Encompass_Preferred
A phospholipid-porphyrin conjugate LNPs containing 1-palmitoyl-2-pyropheophorbide a-sn-glycero-3-PC and encapsulating FAM-labeled siRNA increase cytosolic siRNA levels 1-palmitoyl-2-pyropheophorbide a-sn-glycero-3-PC-stabilized nanoemulsions loaded with paclitaxel and combined with laser irradiation reduce tumor growth and increase survival in a KB squamous cell carcinoma mouse xenograft model
Catalog No. 50-294-5791 Supplier Cayman Chemical Supplier No. 4009141 EA
A phospholipid-porphyrin conjugate LNPs containing 1-palmitoyl-2-pyropheophorbide a-sn-glycero-3-PC and encapsulating FAM-labeled siRNA increase cytosolic siRNA levels 1-palmitoyl-2-pyropheophorbide a-sn-glycero-3-PC-stabilized nanoemulsions loaded with paclitaxel and combined with laser irradiation reduce tumor growth and increase survival in a KB squamous cell carcinoma mouse xenograft model