Learn More
Cayman Chemical IgG1mouse RabbIt MoncLNa 100ug
SDP
Catalog No. 502948549
Encompass_Preferred
Immunogen: Mouse IgG • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG1; (-) Mouse IgG2a, IgG3, IgM, IgE (-) Human, rat IgG • Applications: ELISA, WB • MW = ~150 and 50 kDa for non-reduced and reduced mouse IgG1, respectively
Catalog No. 50-294-8549 Supplier Cayman Chemical Supplier No. 32003100 UG
Immunogen: Mouse IgG • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG1; (-) Mouse IgG2a, IgG3, IgM, IgE (-) Human, rat IgG • Applications: ELISA, WB • MW = ~150 and 50 kDa for non-reduced and reduced mouse IgG1, respectively