Learn More
IgG2 Fc Mouse anti-Human, FITC, Clone: HP6002, Southern Biotech™
Shop All Southern Biotechnology Associates Products

Description
SouthernBiotech monoclonal secondary antibodies offer high sensitivity and low background for use in a variety of applications.
Specifications
Specifications
| Antigen | IgG2 Fc |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | HP6002 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Formulation | Supplied in PBS/NaN3 |
| Host Species | Mouse |
| Immunogen | Human IgG2 myeloma protein |
| Quantity | 0.5 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.