Learn More
Athens Research And Technology Inc IGG2 HU MYELOMA PLASMA 10MG
Supplier:  Athens Research And Technology Inc 16160907072M10MG
Encompass_Preferred
Immunoglobulin G2, Human Myeloma Plasma: 10 MG; Frozen in 20 mM phosphate, pH 7.4, with 150 mM NaCl and 0.05% NaN3; MW 146,000; EC= 1.36; Purity: > =95% by SDS-PAGE. Native; Purified from human myeloma plasma in the Athens Research & Technology laboratory. Immunology, Molecular Biology Reagent
Catalog No. 50-489-807