Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IgG2 ELISA Kit

Catalog No. BMS2093TEN
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMS2093TEN 10 x 96 Tests
BMS2093FIVE 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BMS2093TEN Supplier Invitrogen™ Supplier No. BMS2093TEN
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human ImmunoGlobulin G2 (Hu IgG2) ELISA quantitates Hu IgG2 in human serum or plasma. The assay will exclusively recognize both natural and recombinant Hu IgG2. Principle of the method The Human IgG2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Human IgG2, also known as human immunoglobulin G2, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the second most abundant subclass of IgG in the blood and plays a key role in defense against pathogens. IgG2 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG2 is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG2 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.2-75 ng/mL
Assay Sensitivity 0.25 ng/mL
Content And Storage Aluminum pouch(es) with a Microplate coated with monoclonal antibody to human IgG2
HRP-Conjugate anti-human IgG monoclonal antibody
human IgG2 Standard lyophilized, 150 ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias IGHG2; immunoglobulin heavy constant gamma 2 (G2m marker)IGHG2; immunoglobulin heavy constant gamma 2 (G2m marker);
Interassay CV 7.2%
Intraassay CV 6.2%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 20 μL, Serum 20 μL
Storage Requirements 2-8°C
Target Human IgG2
Test Time 1 hr 20 min
Total Assay Time 1 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.