Learn More
AdipoGen IgG2 human mAb FITC
SDP
Supplier:  AdipoGen AG29B0009FC050
Encompass_Preferred
Monoclonal Antibody. Recognizes human anti-IgG2 Fc domain. Applications FACS, ICC. Isotype Mouse IgG1. Clone 31-7-4. Liquid. In PBS containing 0.02% sodium azide. Detects human IgG2 antibody and human IgG2Fc proteins and can be used as a secondary control antibody. Can also be used for detection and coating/coupling of Fc-IgG2 based proteins on ELISA kit plates.
Catalog No. 50-256-3097