Learn More
Athens Research And Technology Inc IGG2 HUMAN PLASMA 1MG
Supplier:  Athens Research And Technology Inc 161609070721MG
Encompass_Preferred
Immunoglobulin G2, Human Normal Plasma: 1 MG; Frozen in 20 mM phosphate, pH 7.4, with 150 mM NaCl and 0.05% NaN3; MW 150,000; EC= 1.36; Native; Purified from human plasma in the Athens Research & Tech lab. Immunology, Molecular Biology Reagent
Catalog No. 50-489-802