Learn More
Alpco Ltd Mouse IgG2A ELISA
SDP
Supplier:  Alpco Ltd 41I2AMSE01
Encompass
The Mouse IgG2A ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgG2A in biological samples of mice. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3582