Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse IgG2a Fc, Avi Tag (Avitag™);HEK293;500UG;IGA,IgG2A Fc,IgG1 Fc,IgG2,IgG3,IgG4,IgG2a,IgG Fc,IgG1,IgG2a Fc
Catalog No. 501659143
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse IgG2a Fc, Avi Tag (Avitag™);500UG;Biotinylated Mouse IgG Fc, Avi Tag (IGA-M8210) is expressed from human 293 cells (HEK293). It contains AA Glu 98 - Lys 330 (Accession # P01863).;This protein carries an Avi tag (Avitag™) at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 36 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Recombinant Protein A at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse IgG2a Fc (Cat. No. IGA-M8210) with a linear range of 0.4-12.5 ng/mL (QC tested).
Catalog No. 50-165-9143 Supplier ACROBiosystems Supplier No. IGAM8210500UG
Recombinant Protein;Biotinylated Mouse IgG2a Fc, Avi Tag (Avitag™);500UG;Biotinylated Mouse IgG Fc, Avi Tag (IGA-M8210) is expressed from human 293 cells (HEK293). It contains AA Glu 98 - Lys 330 (Accession # P01863).;This protein carries an Avi tag (Avitag™) at the C-terminus. The protein has a calculated MW of 28.2 kDa. The protein migrates as 36 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Recombinant Protein A at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse IgG2a Fc (Cat. No. IGA-M8210) with a linear range of 0.4-12.5 ng/mL (QC tested).