Learn More
Cayman Chemical IgG2amouse RabbIt Monclon 50ug
SDP
Catalog No. 502947206
Encompass_Preferred
Immunogen: Mouse IgG2a • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG2a; (-) Mouse IgG1, IgG2b, IgG2c, IgG3, IgM, IgA, IgE (-) Human, rat IgG • Applications: ELISA, WB (non-reducing conditions) • MW = ~150 kDa
Catalog No. 50-294-7206 Supplier Cayman Chemical Supplier No. 3235250 UG
Immunogen: Mouse IgG2a • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG2a; (-) Mouse IgG1, IgG2b, IgG2c, IgG3, IgM, IgA, IgE (-) Human, rat IgG • Applications: ELISA, WB (non-reducing conditions) • MW = ~150 kDa