Learn More
Enzo Life Sciences IgG2b (mouse), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900110
Encompass_Preferred
The IgG2b (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minutes. Ideal for determining the isotype and concentration of monoclonal antibodies. Sensitivity: 1.05 ng/ml (range 7.81 - 250 ng/ml). Assay Time: 90 minutes. Applications: ELISA, Colorimetric detection . Species reactivity: Mouse .
Catalog No. 50-200-6845