Learn More
Cayman Chemical IgG2bmouse RabbIt Monclon 50ug
SDP
Catalog No. 502947536
Encompass_Preferred
Immunogen: Mouse IgG2b • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG2b; (-) Mouse IgG1, IgG2a, IgG3, IgM, IgA, IgE; (-) Human, goat, rat IgG • Applications: ELISA, WB • MW: ~150 and 50 kDa for non-reduced and reduced mouse IgG1, respectively
Catalog No. 50-294-7536 Supplier Cayman Chemical Supplier No. 3235350 UG
Immunogen: Mouse IgG2b • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG2b; (-) Mouse IgG1, IgG2a, IgG3, IgM, IgA, IgE; (-) Human, goat, rat IgG • Applications: ELISA, WB • MW: ~150 and 50 kDa for non-reduced and reduced mouse IgG1, respectively