Learn More
Cayman Chemical IgG2cmouse RabbIt MoncLN 100ug
SDP
Catalog No. 502948788
Encompass_Preferred
Immunogen: Mouse IgG2c • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (-) Human, rat IgG; (-) Mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgE, IgM • Applications: ELISA, WB (non-reducing conditions)
Catalog No. 50-294-8788 Supplier Cayman Chemical Supplier No. 32102100 UG
Immunogen: Mouse IgG2c • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (-) Human, rat IgG; (-) Mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgE, IgM • Applications: ELISA, WB (non-reducing conditions)