Learn More
Cayman Chemical IgG2cmouse RabbIt Monclon 50ug
SDP
Catalog No. 502946920
Encompass_Preferred
Immunogen: Mouse IgG2c • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG2c; (-) Mouse IgG1, IgG2a, IgG2b, IgG3, IgM, IgA, IgE (-) Human, rat IgG • Applications: ELISA, WB (non-reducing conditions)
Catalog No. 50-294-6920 Supplier Cayman Chemical Supplier No. 3235750 UG
Immunogen: Mouse IgG2c • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgG2c; (-) Mouse IgG1, IgG2a, IgG2b, IgG3, IgM, IgA, IgE (-) Human, rat IgG • Applications: ELISA, WB (non-reducing conditions)