Learn More
Cayman Chemical IgG2humn RabbIt Monclona 100ug
SDP
Catalog No. 502947197
Encompass_Preferred
Immunogen: Human IgG2 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgG2 Heavy Chain; (-) Human IgA, IgD, IgE, IgG1, IgG3, IgG4, IgM; (-) Cynomolgus monkey, goat, mouse, rat, rhesus monkey IgG • Application: ELISA
Catalog No. 50-294-7197 Supplier Cayman Chemical Supplier No. 32121100 UG
Immunogen: Human IgG2 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgG2 Heavy Chain; (-) Human IgA, IgD, IgE, IgG1, IgG3, IgG4, IgM; (-) Cynomolgus monkey, goat, mouse, rat, rhesus monkey IgG • Application: ELISA