Learn More
Cayman Chemical IgG2humn RabbIt Monclona 50ug
SDP
Catalog No. 502950891
Encompass_Preferred
Immunogen: Human IgG2 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgG2; (-) Human IgG1, IgG3, IgG4, IgM, IgA, IgD, IgE; (-) Goat, mouse, rat IgG • Applications: ELISA
Catalog No. 50-295-0891 Supplier Cayman Chemical Supplier No. 3237450 UG
Immunogen: Human IgG2 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgG2; (-) Human IgG1, IgG3, IgG4, IgM, IgA, IgD, IgE; (-) Goat, mouse, rat IgG • Applications: ELISA