Learn More
Cayman Chemical IgG3humn RabbIt Monclona 100ug
SDP
Catalog No. 502948192
Encompass_Preferred
Immunogen: Human IgG3 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (-) Human IgG1, IgG2, IgG4, IgM, IgA, IgD, IgE; (-) Cynomolgus monkey, goat, mouse, rat, or rhesus monkey IgG • Applications: ELISA, ICC, IHC
Catalog No. 50-294-8192 Supplier Cayman Chemical Supplier No. 32122100 UG
Immunogen: Human IgG3 • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (-) Human IgG1, IgG2, IgG4, IgM, IgA, IgD, IgE; (-) Cynomolgus monkey, goat, mouse, rat, or rhesus monkey IgG • Applications: ELISA, ICC, IHC