Learn More
Cayman Chemical IgG3mouse RabbIt MoncLNa 100ug
SDP
Catalog No. 502949648
Encompass_Preferred
Immunogen: Mouse IgG3 • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (-) Mouse IgG1, IgG2a, IgG2b, IgM, IgA, IgE (-) Human and rat IgG • Applications: ELISA, WB • MW = None listed
Catalog No. 50-294-9648 Supplier Cayman Chemical Supplier No. 32098100 UG
Immunogen: Mouse IgG3 • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (-) Mouse IgG1, IgG2a, IgG2b, IgM, IgA, IgE (-) Human and rat IgG • Applications: ELISA, WB • MW = None listed